Methyl-4-amino-3-iodbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-3-iodobenzoic acid methyl ester, Methyl 4-amino-3-iodobenzoate

M-Clarity™ Qualitätsstufe = MQ100

515183-25G 515183-5G
SIAL515183-25GEA 300 EUR
SIAL515183-25G SIAL515183-5G
Methyl-4-amino-3-iodbenzoat, Sigma-Aldrich®
Methyl-4-amino-3-iodbenzoat
Formel: C₈H₈INO₂
Molecular Weight: 277,0600 g/mol
MDL: MFCD00090423
CAS-Nummer: 19718-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR