Methyl-2-amino-3-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-amino-3-methylbenzoate

CA-4452-5G CA-4452-1G
COBBCA-4452-5GEA 30.8 EUR
COBBCA-4452-5G COBBCA-4452-1G
Methyl-2-amino-3-methylbenzoat
Methyl-2-amino-3-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD06200918
CAS-Nummer: 22223-49-0
EINECS: 244-847-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR