Methyl-3-amino-4-methylbenzoat
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-amino-4-methylbenzoate
BTB03382.250
BTB03382.1
MAYBBTB03382.250EA
0
EUR
MAYBBTB03382.250
MAYBBTB03382.1
Methyl-3-amino-4-methylbenzoat
Methyl-3-amino-4-methylbenzoat
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol |
MDL:
MFCD00025206 CAS-Nummer: 18595-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...