Methyl-4-amino-3-methylbenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4-amino-3-methylbenzoate

11X-0812.1 11X-0812.10 11X-0812.5 11X-0812.500
BIOT11X-0812.1EA 0 EUR
BIOT11X-0812.1 BIOT11X-0812.10 BIOT11X-0812.5 BIOT11X-0812.500
Methyl-4-amino-3-methylbenzoat
Methyl-4-amino-3-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
MDL: MFCD00102230
CAS-Nummer: 18595-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR