Methyl-4-amino-3-methylbenzoat

Lieferant: FLUOROCHEM

Synonyme: Methyl 4-amino-3-methylbenzoate

CFF397.10 CFF397.25 017420-10G 017420-25G 017420-1G
CHMPCFF397.10EA 455 EUR
CHMPCFF397.10 CHMPCFF397.25 FLUO017420-10G FLUO017420-25G FLUO017420-1G
Methyl-4-amino-3-methylbenzoat
Methyl-4-amino-3-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD00102230
CAS-Nummer: 18595-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR