3-(2-Bromphenyl)-5-ethyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6282-5G HC-6282-1G
COBBHC-6282-5GEA 451 EUR
COBBHC-6282-5G COBBHC-6282-1G
3-(2-Bromphenyl)-5-ethyl-1,2,4-oxadiazol
3-(2-Bromophenyl)-5-ethyl-1,2,4-oxadiazole
Formel: C₁₀H₉BrN₂O
MDL: MFCD11504917
CAS-Nummer: 1072944-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR