3-(3-Bromphenyl)-5-ethyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-2516-5G HC-2516-1G
COBBHC-2516-5GEA 460 EUR
COBBHC-2516-5G COBBHC-2516-1G
3-(3-Bromphenyl)-5-ethyl-1,2,4-oxadiazol
3-(3-Bromophenyl)-5-ethyl-1,2,4-oxadiazole
Formel: C₁₀H₉BrN₂O
MDL: MFCD12026022
CAS-Nummer: 911049-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR