3-(3-Bromphenyl)-5-methyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-2279-5G HC-2279-1G
COBBHC-2279-5GEA 385 EUR
COBBHC-2279-5G COBBHC-2279-1G
3-(3-Bromphenyl)-5-methyl-1,2,4-oxadiazol
3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole
Formel: C₉H₇BrN₂O
MDL: MFCD08087563
CAS-Nummer: 160377-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR