3-(4-Bromphenyl)-5-tert-butyl-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS
HC-6436-1G
HC-6436-5G
COBBHC-6436-1GEA
149
EUR
COBBHC-6436-1G
COBBHC-6436-5G
3-(4-Bromphenyl)-5-tert-butyl-1,2,4-oxadiazol
3-(4-Bromphenyl)-5-tert-butyl-1,2,4-oxadiazol
Formel:
C₁₂H₁₃BrN₂O |
MDL:
MFCD02310125 CAS-Nummer: 676131-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...