3-(4-Bromphenyl)-5-ethyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6434-5G HC-6434-1G
COBBHC-6434-5GEA 375 EUR
COBBHC-6434-5G COBBHC-6434-1G
3-(4-Bromphenyl)-5-ethyl-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-ethyl-1,2,4-oxadiazole
Formel: C₁₀H₉BrN₂O
MDL: MFCD00635507
CAS-Nummer: 364743-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR