3-(4-Bromphenyl)-5-methyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

OT-0436-5G OT-0436-1G
COBBOT-0436-5GEA 312 EUR
COBBOT-0436-5G COBBOT-0436-1G
3-(4-Bromphenyl)-5-methyl-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole
Formel: C₉H₇BrN₂O
MDL: MFCD00487341
CAS-Nummer: 118183-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR