3-(Benzyloxy)-4,5-dimethoxybenzaldehyd

Lieferant: Thermo Scientific

BTB13918.250 BTB13918.1
MAYBBTB13918.250EA 0 EUR
MAYBBTB13918.250 MAYBBTB13918.1
3-(Benzyloxy)-4,5-dimethoxybenzaldehyd
3-(Benzyloxy)-4,5-dimethoxybenzaldehyd
Formel: C₁₆H₁₆O₄
CAS-Nummer: 57230-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR