2-Isopropoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5497-5G BB-5497-1G
COBBBB-5497-5GEA 0 EUR
COBBBB-5497-5G COBBBB-5497-1G
2-Isopropoxy-3-pyridinboronsäure
2-Isopropoxy-3-pyridinboronsäure
Formel: C₈H₁₂BNO₃
MDL: MFCD11855840
CAS-Nummer: 1150114-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR