2-Propoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Propoxypyridine-3-boronic acid, (2-Propoxypyridin-3-yl)boronic acid

BB-8894-5G BB-8894-1G
COBBBB-8894-5GEA 884 EUR
COBBBB-8894-5G COBBBB-8894-1G
2-Propoxy-3-pyridinboronsäure
2-Propoxy-3-pyridineboronic acid
Formel: C₈H₁₂BNO₃
Molecular Weight: 181 g/mol
MDL: MFCD13195680
CAS-Nummer: 1218790-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR