3-Methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methoxypyridine-4-boronic acid

BB-5578-5G BB-5578-1G BB-5578-250M
COBBBB-5578-5GEA 1590 EUR
COBBBB-5578-5G COBBBB-5578-1G COBBBB-5578-250M
3-Methoxy-4-pyridinboronsäure
3-Methoxy-4-pyridineboronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152.9400 g/mol
MDL: MFCD06801698
CAS-Nummer: 1008506-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR