3-Methoxy-4-pyridinboronsäure Hydrat
Lieferant: COMBI-BLOCKS
BB-8536-5G
BB-8536-1G
COBBBB-8536-5GEA
1440
EUR
COBBBB-8536-5G
COBBBB-8536-1G
3-Methoxy-4-pyridinboronsäure Hydrat
3-Methoxy-4-pyridinboronsäure Hydrat
Formel:
C₆H₈BNO₃·H₂O |
MDL:
MFCD07781246 CAS-Nummer: 1072952-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...