4-Methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methoxypyridin-3-yl)boronic acid

BB-5108-5G BB-5108-1G BB-5108-250M
COBBBB-5108-5GEA 2350 EUR
COBBBB-5108-5G COBBBB-5108-1G COBBBB-5108-250M
4-Methoxy-3-pyridinboronsäure
4-Methoxy-3-pyridineboronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152.9436 g/mol
MDL: MFCD01114676
CAS-Nummer: 355004-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR