4-Methoxy-3-pyridinboronsäure Hydrat
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Methoxypyridin-3-yl)boronic acid hydrate, 4-Methoxypyridine-3-boronic acid hydrate
BB-8509-5G
BB-8509-1G
COBBBB-8509-5GEA
667
EUR
COBBBB-8509-5G
COBBBB-8509-1G
4-Methoxy-3-pyridinboronsäure Hydrat
4-Methoxy-3-pyridinboronsäure Hydrat
Formel:
C₆H₈BNO₃·H₂O Molecular Weight: 152.94 g/mol (anhy) |
MDL:
MFCD07186213 CAS-Nummer: 1256355-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...