5-Methoxy-2-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5882-1G BB-5882-250M
COBBBB-5882-1GEA 0 EUR
COBBBB-5882-1G COBBBB-5882-250M
5-Methoxy-2-pyridinboronsäure
5-Methoxy-2-pyridineboronic acid
Formel: C₆H₈BNO₃
MDL: MFCD08752653
CAS-Nummer: 1142944-78-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR