2-Isopropoxy-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Isopropoxypyridin-3-yl)boronic acid, 2-Isoproxypyridine-5-boronic acid

BB-8520-5G BB-8520-1G
COBBBB-8520-5GEA 685 EUR
COBBBB-8520-5G COBBBB-8520-1G
2-Isopropoxy-5-pyridinboronsäure
2-Isopropoxy-5-pyridinboronsäure
Formel: C₈H₁₂BNO₃
Molecular Weight: 181,00 g/mol
MDL: MFCD07368870
CAS-Nummer: 870521-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR