4-Methoxy-3-pyridinboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-5210-1G BB-5210-5G
COBBBB-5210-1GEA 0 EUR
COBBBB-5210-1G COBBBB-5210-5G
4-Methoxy-3-pyridinboronsäure Hydrochlorid
4-Methoxy-3-pyridinboronsäure Hydrochlorid
Formel: C₆H₉BClNO₃
MDL: MFCD04113800
CAS-Nummer: 874959-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR