5-Fluor-2-methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-methoxypyridine-4-boronic acid

BB-5259-1G BB-5259-250M
COBBBB-5259-1GEA 183 EUR
COBBBB-5259-1G COBBBB-5259-250M
5-Fluor-2-methoxy-4-pyridinboronsäure
5-Fluoro-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BFNO₃
Molecular Weight: 170.9300 g/mol
MDL: MFCD06657887
CAS-Nummer: 1043869-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR