3-Fluor-2-methoxy-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Fluoro-6-methoxypyridin-3-yl)boronic acid, 3-Fluoro-2-methoxypyridine-5-boronic acid

BB-8460-1G BB-8460-250M
COBBBB-8460-1GEA 640 EUR
COBBBB-8460-1G COBBBB-8460-250M
3-Fluor-2-methoxy-5-pyridinboronsäure
3-Fluor-2-methoxy-5-pyridinboronsäure
Formel: C₆H₇BFNO₃
Molecular Weight: 170.9341 g/mol
MDL: MFCD04112520
CAS-Nummer: 856250-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR