4-Methyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methylpyridin-3-yl)boronic acid, 4-Methylpyridine-3-boronic acid

BB-3920-5G BB-3920-1G
COBBBB-3920-5GEA 336 EUR
COBBBB-3920-5G COBBBB-3920-1G
4-Methyl-3-pyridinboronsäure
4-Methyl-3-pyridineboronic acid
Formel: C₆H₈BNO₂
Molecular Weight: 136.94 g/mol
MDL: MFCD03788244
CAS-Nummer: 148546-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR