5-Methyl-2-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Methylpyridin-2-yl)boronic acid

BB-2393-1G BB-2393-5G BB-2393-250M
COBBBB-2393-1GEA 734 EUR
COBBBB-2393-1G COBBBB-2393-5G COBBBB-2393-250M
5-Methyl-2-pyridinboronsäure
5-Methyl-2-pyridineboronic acid
Formel: C₆H₈BNO₂
Molecular Weight: 136,95 g/mol
MDL: MFCD00674179
CAS-Nummer: 372963-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR