5-Methyl-3-pyridinboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Methylpyridine-3-boronic acid
H52891.MD
H52891.03
H52891.MDEA
31.1
EUR
H52891.MD
H52891.03
5-Methyl-3-pyridinboronsäure ≥98%
5-Methyl-3-pyridineboronic acid
Purity: 98%
CAS No.: 173999-18-3
Molecular Formula: C6H8BNO2
Formula Weight: 136.95
Storage Temperature: -30°C to -10°C
Melting Point:
Formel:
C₆H₈BNO₂ Molecular Weight: 136,95 g/mol Schmelzpunkt: >300 °C |
MDL:
MFCD03428528 CAS-Nummer: 173999-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...