5-Methyl-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Methylpyridine-3-boronic acid
BB-3596-5G
BB-3596-1G
COBBBB-3596-5GEA
587
EUR
COBBBB-3596-5G
COBBBB-3596-1G
5-Methyl-3-pyridinboronsäure
5-Methyl-3-pyridineboronic acid
Formel:
C₆H₈BNO₂ Molecular Weight: 136.95 g/mol |
MDL:
MFCD03428528 CAS-Nummer: 173999-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...