2-(Trifluormethyl)-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Trifluoromethylpyridine-5-boronic acid pinacol ester, 2-(Trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester
PN-8525-1G
PN-8525-5G
COBBPN-8525-1GEA
279
EUR
COBBPN-8525-1G
COBBPN-8525-5G
2-(Trifluormethyl)-5-pyridinboronsäurepinakolester
2-(Trifluormethyl)-5-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₅BF₃NO₂ Molecular Weight: 273.592 g/mol |
MDL:
MFCD11226774 CAS-Nummer: 1218790-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...