2-Isobutoxy-5-(trifluormethyl)-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6238-1G PN-6238-5G PN-6238-250M
COBBPN-6238-1GEA 667 EUR
COBBPN-6238-1G COBBPN-6238-5G COBBPN-6238-250M
2-Isobutoxy-5-(trifluormethyl)-3-pyridinboronsäurepinakolester
2-Isobutoxy-5-(trifluormethyl)-3-pyridinboronsäurepinakolester
Formel: C₁₆H₂₃BF₃NO₃
MDL: MFCD16295110
CAS-Nummer: 1256359-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR