2-Fluor-6-methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8412-5G BB-8412-1G
COBBBB-8412-5GEA 0 EUR
COBBBB-8412-5G COBBBB-8412-1G
2-Fluor-6-methyl-5-pyridinboronsäure
2-Fluor-6-methyl-5-pyridinboronsäure
Formel: C₆H₇BFNO₂
MDL: MFCD03411561
CAS-Nummer: 904326-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR