2-Isopropoxy-5-(trifluormethyl)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Isopropoxy-5-(trifluoromethyl)pyridin-3-yl)boronic acid
BB-6237-5G
BB-6237-1G
COBBBB-6237-5GEA
692
EUR
COBBBB-6237-5G
COBBBB-6237-1G
2-Isopropoxy-5-(trifluormethyl)-3-pyridinboronsäure
2-Isopropoxy-5-(trifluormethyl)-3-pyridinboronsäure
Formel:
C₉H₁₁BF₃NO₃ Molecular Weight: 248,9948 g/mol |
MDL:
MFCD15143471 CAS-Nummer: 1218790-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...