2-Methoxy-5-(trifluormethyl)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
BB-5261-1G
BB-5261-5G
COBBBB-5261-1GEA
0
EUR
COBBBB-5261-1G
COBBBB-5261-5G
2-Methoxy-5-(trifluormethyl)-3-pyridinboronsäure
2-Methoxy-5-(trifluoromethyl)-3-pyridineboronic acid
Formel:
C₇H₇BF₃NO₃ |
MDL:
MFCD10696251 CAS-Nummer: 1072946-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...