2-Propoxy-5-(trifluormethyl)-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-6233-5G BB-6233-25G BB-6233-1G
COBBBB-6233-5GEA 451 EUR
COBBBB-6233-5G COBBBB-6233-25G COBBBB-6233-1G
2-Propoxy-5-(trifluormethyl)-3-pyridinboronsäure
2-Propoxy-5-(trifluoromethyl)-3-pyridineboronic acid
Formel: C₉H₁₁BF₃NO₃
MDL: MFCD15143467
CAS-Nummer: 1218790-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR