(2-Bromphenyl)essigsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2-Bromophenyl)acetic acid

CA-4346-25G CA-4346-100G
COBBCA-4346-25GEA 60.9 EUR
COBBCA-4346-25G COBBCA-4346-100G
(2-Bromphenyl)essigsäure
(2-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215.05 g/mol
Siedepunkt: 308
Schmelzpunkt: 107,5
MDL: MFCD00004314
CAS-Nummer: 18698-97-0
EINECS: 242-509-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR