4-Nitrothioanisol

Lieferant: Thermo Scientific

Synonyme: 1-Nitro-4-(methylthio)benzol, 4-Nitrophenylmethylsulfid

RH00769.1 RH00769.10 RH00769.50
MAYBRH00769.1EA 0 EUR
MAYBRH00769.1 MAYBRH00769.10 MAYBRH00769.50
4-Nitrothioanisol
4-Nitrothioanisol
Formel: C₇H₇NO₂S
Molecular Weight: 169,2 g/mol
Siedepunkt: 135…140 °C (2 mmHg)
Schmelzpunkt: 66…68 °C
MDL: MFCD00010868
CAS-Nummer: 701-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR