4-((Trifluormethyl)thio)benzoesäure

Lieferant: COMBI-BLOCKS

OS-7539-25G OS-7539-100G OS-7539-5G
COBBOS-7539-25GEA 0 EUR
COBBOS-7539-25G COBBOS-7539-100G COBBOS-7539-5G
4-((Trifluormethyl)thio)benzoesäure
4-((Trifluormethyl)thio)benzoesäure
Formel: C₈H₅F₃O₂S
MDL: MFCD00040906
CAS-Nummer: 330-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR