3-Chlor-2-fluor-5-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
005681-5G
005681-1G
FLUO005681-5GEA
249
EUR
FLUO005681-5G
FLUO005681-1G
3-Chlor-2-fluor-5-(trifluormethyl)benzoylchlorid
3-Chlor-2-fluor-5-(trifluormethyl)benzoylchlorid
Formel:
C₈H₂Cl₂F₄O Molecular Weight: 261 g/mol Siedepunkt: 232 Schmelzpunkt: 85 |
MDL:
MFCD01631511 CAS-Nummer: 261763-03-5 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...