3-Chlor-2-fluor-6-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
005682-5G
005682-1G
FLUO005682-5GEA
0
EUR
FLUO005682-5G
FLUO005682-1G
3-Chlor-2-fluor-6-(trifluormethyl)benzoylchlorid
3-Chlor-2-fluor-6-(trifluormethyl)benzoylchlorid
Formel:
C₈H₂Cl₂F₄O Siedepunkt: 232 °C (1013 hPa) Schmelzpunkt: 85 °C |
MDL:
MFCD01631344 CAS-Nummer: 186517-45-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...