2-Fluornicotinonitril
Lieferant: COMBI-BLOCKS
Synonyme:
3-Cyan-2-fluorpyridin, 2-Fluor-3-pyridincarbonsäurenitril
OS-7605-5G
OS-7605-25G
OS-7605-1G
OS-7605-100G
COBBOS-7605-5GEA
50.8
EUR
COBBOS-7605-5G
COBBOS-7605-25G
COBBOS-7605-1G
COBBOS-7605-100G
2-Fluornicotinonitril
2-Fluornicotinonitril
Formel:
C₆H₃FN₂ Molecular Weight: 122,1 g/mol Siedepunkt: 196 °C (1013 hPa) Schmelzpunkt: 30…30,5 °C |
MDL:
MFCD03095082 CAS-Nummer: 3939-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...