2-Fluorisonicotinonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluor-4-pyridincarbonsäurenitril, 4-Cyan-2-fluorpyridin

PY-7061-100G PY-7061-1G PY-7061-25G PY-7061-5G
COBBPY-7061-100GEA 724 EUR
COBBPY-7061-100G COBBPY-7061-1G COBBPY-7061-25G COBBPY-7061-5G
2-Fluorisonicotinonitril
2-Fluorisonicotinonitril
Formel: C₆H₃FN₂
Molecular Weight: 122.1 g/mol
MDL: MFCD09027299
CAS-Nummer: 3939-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR