2-Chlor-6-nitrobenzonitril
Lieferant: CHEMPUR
Synonyme:
6-Chloro-2-nitrobenzonitrile
TL00403.250
TL00403.1
CHMPTL00403.250EA
0
EUR
CHMPTL00403.250
MAYBTL00403.1
2-Chlor-6-nitrobenzonitril
2-Chlor-6-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,57 g/mol |
MDL:
MFCD00007203 CAS-Nummer: 6575-07-1 EINECS: 229-496-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...