7-Isochinolinol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
7-Hydroxyisoquinoline, Isoquinolin-7-ol
HC-6002-5G
HC-6002-1G
COBBHC-6002-5GEA
407
EUR
COBBHC-6002-5G
COBBHC-6002-1G
7-Isochinolinol
7-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol |
MDL:
MFCD00456131 CAS-Nummer: 7651-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...