Deblock trichloroacetic acid solution 3% (w/w) in Dichlormethan
Lieferant: Thermo Fisher Scientific
Gefahr
J62821.AH J62821.AO J62821.K4
J62821.AHEA 45.8 EUR
J62821.AH J62821.AO J62821.K4
Deblock trichloroacetic acid solution 3% (w/w) in Dichlormethan
Deblock trichloroacetic acid solution
Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
Dichte: 1,325
Lagertemperatur: Raumtemperatur
MDL: MFCD00004177
EINECS: 200-927-2
UN: 2810
ADR: 6.1,III