4-Brom-N-phenylbenzamid

Lieferant: COMBI-BLOCKS

CA-4415-5G CA-4415-25G CA-4415-1G
COBBCA-4415-5GEA 404 EUR
COBBCA-4415-5G COBBCA-4415-25G COBBCA-4415-1G
4-Brom-N-phenylbenzamid
4-Brom-N-phenylbenzamid
Formel: C₁₃H₁₀BrNO
Molecular Weight: 276,13 g/mol
MDL: MFCD00624477
CAS-Nummer: 6846-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR