4-Thiazolcarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Thiazole-4-carboxylic acid
HI-1121-5G
HI-1121-25G
HI-1121-1G
COBBHI-1121-5GEA
63.9
EUR
COBBHI-1121-5G
COBBHI-1121-25G
COBBHI-1121-1G
4-Thiazolcarbonsäure
4-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S Molecular Weight: 129,1371 g/mol |
MDL:
MFCD00623589 CAS-Nummer: 3973-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...