3-Brom-N,N-diethyl-5-(trifluormethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-5-(N,N-diethylsulphamoyl)benzotrifluoride
HC-6224-5G
HC-6224-1G
COBBHC-6224-5GEA
451
EUR
COBBHC-6224-5G
COBBHC-6224-1G
3-Brom-N,N-diethyl-5-(trifluormethyl)benzolsulfonamid
3-Brom-N,N-diethyl-5-(trifluormethyl)benzolsulfonamid
Formel:
C₁₁H₁₃BrF₃NO₂S |
MDL:
MFCD09800988 CAS-Nummer: 951885-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...