3-Brom-N-ethyl-5-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-(N-ethylsulphamoyl)benzotrifluoride

HC-6293-1G HC-6293-5G
COBBHC-6293-1GEA 0 EUR
COBBHC-6293-1G COBBHC-6293-5G
3-Brom-N-ethyl-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-ethyl-5-(trifluormethyl)benzolsulfonamid
Formel: C₉H₉BrF₃NO₂S
MDL: MFCD09801005
CAS-Nummer: 951884-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR