3-Brom-N-propyl-5-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-(N-propylsulphamoyl)benzotrifluoride

HC-6311-5G HC-6311-1G
COBBHC-6311-5GEA 595 EUR
COBBHC-6311-5G COBBHC-6311-1G
3-Brom-N-propyl-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-propyl-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₀H₁₁BrF₃NO₂S
MDL: MFCD09801014
CAS-Nummer: 951884-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR