3-Brom-N-cyclopropyl-5-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-(N-cyclopropylsulphamoyl)benzotrifluoride

HC-6312-5G HC-6312-1G
COBBHC-6312-5GEA 0 EUR
COBBHC-6312-5G COBBHC-6312-1G
3-Brom-N-cyclopropyl-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-cyclopropyl-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₀H₉BrF₃NO₂S
MDL: MFCD09801015
CAS-Nummer: 951884-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR