3-Brom-N-(3-methoxypropyl)-5-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-[N-(3-methoxypropyl)sulphamoyl]benzotrifluoride

HC-6305-1G HC-6305-5G
COBBHC-6305-1GEA 0 EUR
COBBHC-6305-1G COBBHC-6305-5G
3-Brom-N-(3-methoxypropyl)-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-(3-methoxypropyl)-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₁H₁₃BrF₃NO₃S
MDL: MFCD09801009
CAS-Nummer: 951884-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR